Learn More
Miele Professional™ ProCare Lab 10 AP
Product Code. 32237211 Shop All Miele Products
Change view
Click to view available options
Quantity:
5 L
Unit Size:
5 L
Product Code. Quantity Unit Size
32237211 5 L 5 L
1 options
This item is not returnable. View return policy
Product Code. 32237211 Supplier Miele Professional™ Supplier No. 12958900
  • ONLINE EXCLUSIVE PRICE
    77.05€
    48.00€ / 5 litros
    Ahorro 29.05€
    37% Descuento
 Restricted Item
Fecha estimada de envoi: 24-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For machine-based universal cleaning of laboratory glassware and utensils.
  • Alkaline cleaning agent with a high percentage of dispersants
  • Free from surfactants, phosphates and oxidants
  • Specially developed for Miele Professional washer-disinfectors
  • Suitable for stainless steel, laboratory glassware, ceramics and plastics
  • For cleaning stubborn or dried-on residues
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleaning Agent
Color-coded Yes
Form Liquid
Scent Unscented
Sterility Non-sterile
Quantity 5 L