Learn More
Pro-Lab Diagnostics™ Prolex™ Streptococcal Grouping Latex Kits
Product Code. 11959044
Change view
Click to view available options
Quantity:
3 mL
Type:
Group A Latex Reagent
Group B Latex Reagent
Group C Latex Reagent
Group D Latex Reagent
Group F Latex Reagent
Group G Latex Reagent
Unit Size:
Pack of 60
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Type Unit Size
11959044 3 mL Group G Latex Reagent Pack of 60
11909044 3 mL Group A Latex Reagent Pack of 60
11929044 3 mL Group C Latex Reagent Pack of 60
11949044 3 mL Group F Latex Reagent Pack of 60
11939044 3 mL Group D Latex Reagent Pack of 60
11919044 3 mL Group B Latex Reagent Pack of 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11959044 Supplier Pro-Lab Diagnostics™ Supplier No. PL.036
 Restricted Item
Fecha estimada de envoi: 07-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
For grouping Lancefield betahaemolytic Streptococci
  • Instant results obtained at room temperature
  • Advanced extraction process using modified nitrous acid reagents
  • Enhanced results with blue carboxylate-modified microparticles (CM-MP)
  • As few as one colony required
  • Colour coded reagents and vials for ease of selection
  • Available individually
Specifications
Content And Storage 2°C to 8°C
Type Group G Latex Reagent
Includes 3mL Blue Latex particles coated with purified Rabbit Antibodies
Quantity 3 mL