missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PRKD1 (PKC mu), GST Tag Recombinant Protein
Human PRKD1 (PKC mu), GST Tag Recombinant Protein
Marca: Invitrogen PV3791
Detalles adicionales : Peso : 0.05250kg
Descripción
PRKC1 (PKC mu) is a serine⁄threonine protein kinase that is phospholipid-dependant and activated by diacylglycerol and phorbol ester TPA. PRKC1 behaves similarly to most PKC isoenzymes of the PKC family, however, PRKC1 differs in some structural and enzymatic features from the other PKC isotypes, and has been shown to be involved in the signal transduction of a wide range of biological responses.Especificaciones
NP_002733 | |
114294, 5587 | |
10 μg | |
PKD Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: CAMK Group | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
protein kinase D1 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
129.3 kDa | |
PRKD1 (PKC mu), PKD | |
Human | |
GST Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
PRKD1 | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |