missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKCZ (PKC zeta) Recombinant Human Protein
PRKCZ (PKC zeta) Recombinant Human Protein
Marca: Invitrogen P2268
Detalles adicionales : Peso : 3.36000kg
Descripción
The PKC family of serine⁄threonine kinases, including PRKCZ (PKC zeta), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent.Especificaciones
NP_002735 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
67.7kDa | |
20μg | |
For research use only. Not for use in diagnostic procedures | |
PRKCZ (PKC zeta) | |
PRKCZ was pre-diluted in enzyme dilution buffer and assayed in 20mM HEPES (pH 7.4), 10mM MgCl2, 0.1mM EGTA, 1X lipid mix (200μg/mL Phosphatidyl Serine, 20μg/mL Diacylglycerol, 1mM HEPES (pH 7.4), 0.03% CHAPS), 500μM ATP 200μg/mL Histone H1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C | |
PRKCZ (PKC zeta) Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.53mg/mL | |
5584, 5590 | |
PRKCZ (PKC zeta) Recombinant Human Protein | |
–80°C | |
1 Each | |
PRKCZ (PKC zeta) Recombinant Human Protein | |
PKC Kinase Family | |
1,100nmoles of phosphate transferred to PKC epsilon peptide substrate (ERMRPRKRQGSVRRRV) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.5 to 1μg/mL | |
None | |
Kinases and inhibitors | |
75% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.