Learn More
PRKCG (PKC gamma) Recombinant Human Protein
PRKCG (PKC gamma) Protein
Marca:  Invitrogen P2233
Detalles adicionales : Peso : 3.18000kg
 Ver más versiones de este producto
Código de producto. 10443822
  • 381.00€ / cada uno
Fecha de envío estimada
del proveedor 21-08-2019
Añadir a la cesta
Descripción
Descripción
The PKC family of serine⁄threonine kinases, including PRKCG (PKC gamma), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent.
Especificaciones
Especificaciones
NP_002730
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
78.4kDa
95% by SDS-PAGE
–80°C
1 Each
PRKCG (PKC gamma)
PRKCG (PKC gamma) was pre-diluted in enzyme dilution buffer and assayed in 20mM HEPES (pH 7.4), 10mM MgCl2, 100μM CaCl2, 1X lipid mix (100μg/mL phosphatidylserine, 20μg/mL diacylglycerol, 1mM HEPES (pH 7.4), 0.03% CHAPS), 500μM ATP 200μg/mL histone H1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
PKC
1800nmoles of phosphate transferred to histone H1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.5 to 2μg/mL
None
0.4mg/mL
5582
PRKCG (PKC gamma) Recombinant Human Protein
5μg
For research use only. Not for use in diagnostic procedures
Serine/threonine kinase
PRKCG (PKC gamma) Recombinant Human Protein
Kinase assay
Baculovirus
PRKCG (PKC gamma) Recombinant Human Protein
Human
Videos
Documentos
For Research Use Only. Not for use in diagnostic procedures.