missing translation for 'onlineSavingsMsg'
Learn More

PRKCB1 (PKC beta I) Recombinant Human Protein

PRKCB1 (PKC beta I)

395.00€ - 5820.00€

Especificaciones

Accession Number NP_991100.1
Ensayos PRKCB1 (PKC beta I) was pre-diluted in enzyme dilution buffer and assayed in 20mM HEPES (pH 7.4), 10mM MgCl2, 100μM CaCl2, 1X lipid mix (100μg/mL phosphatidylserine, 20μg/mL diacylglycerol, 1mM HEPES (pH 7.4), 0.03% CHAPS), 500μM ATP 200μg/mL histone H1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Nombre común PRKCB1 (PKC beta I) Recombinant Human Protein
Expression System Baculovirus
Familia de cinasas PKC
Ver más especificaciones

Productos 3
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
11559236 Ver documentos Invitrogen
P2291
5μg 395.00€
5 microgramos Ver productos alternativos
Ahorre hasta 
»
Fecha de envío estimada
del proveedor 26-06-2020
Añadir a la cesta
 
11529236 Ver documentos Invitrogen
P2281
20μg 640.00€
20 microgramos Ver productos alternativos
Ahorre hasta 
»
Fecha de envío estimada
del proveedor 26-06-2020
Añadir a la cesta
 
10413062 Ver documentos Invitrogen
PV3601
1mg 5820.00€
1 miligramo Ver productos alternativos
Ahorre hasta 
»
Fecha de envío estimada
del proveedor 11-06-2020
Añadir a la cesta
 
Descripción

Descripción

The PKC family of serine⁄threonine kinases, including PRKCB1 (PKC beta I), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent.
Especificaciones

Especificaciones

NP_991100.1
PRKCB1 (PKC beta I) Recombinant Human Protein
PKC
PRKCB1 (PKC beta I) Recombinant Human Protein
PRKCB1 (PKC beta I) Recombinant Human Protein
For research use only. Not for use in diagnostic procedures
Kinase assay
None
PRKCB1 (PKC beta I) was pre-diluted in enzyme dilution buffer and assayed in 20mM HEPES (pH 7.4), 10mM MgCl2, 100μM CaCl2, 1X lipid mix (100μg/mL phosphatidylserine, 20μg/mL diacylglycerol, 1mM HEPES (pH 7.4), 0.03% CHAPS), 500μM ATP 200μg/mL histone H1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Baculovirus
76.9kDa
1 Each
95% by SDS-PAGE
–80°C
Human

For Research Use Only. Not for use in diagnostic procedures.