Learn More
PRKCB1 (PKC beta I) Recombinant Human Protein
PRKCB1 (PKC beta I)
Marca:  Invitrogen PV3601
Detalles adicionales : Peso : 0.05250kg
 Ver más versiones de este producto
Código de producto. 10413062
  • 5820.00€ / 1 miligramo
Fecha de envío estimada
del proveedor 08-06-2020
Añadir a la cesta
Descripción
Descripción
The PKC family of serine⁄threonine kinases, including PRKCB1 (PKC beta I), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent.
Especificaciones
Especificaciones
NP_991100.1
Kinase assay
Protein assays and analysis
Frozen
PRKCB1 (PKC beta I) Recombinant Human Protein
1mg
For research use only. Not for use in diagnostic procedures
PKCB, PRKCB2, PKC-beta, PRKCB1, PKC beta I
PRKCB1 (PKC beta I) was pre-diluted in enzyme dilution buffer and assayed in 20mM HEPES (pH 7.4), 10mM MgCl2, 100μM CaCl2, 1X lipid mix (100μg/mL phosphatidylserine, 20μg/mL diacylglycerol, 1mM HEPES (pH 7.4), 0.03% CHAPS), 500μM ATP 200μg/mL histone H1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
PRKCB1 (PKC beta I) Recombinant Human Protein
Human
Baculovirus
0.75mg/mL
Frozen
76.9kDa
95% by SDS-PAGE
–80°C
1 Each
PRKCB1 (PKC beta I) Recombinant Human Protein
PKC
1,300nmoles of phosphate transferred to histone H1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.34 to 0.67μg/mL
None
Videos
Documentos
For Research Use Only. Not for use in diagnostic procedures.