missing translation for 'onlineSavingsMsg'
Learn More

PRKAR2A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

140.00€ - 399.00€

Especificaciones

Antígeno PRKAR2A
Concentración 0.31 mg/mL
Aplicaciones Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
16826333
Ver documentos Detalles de la promoción
Proteintech
10142-2-AP-20UL
20 μL
140.00€
20 microlitros
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 17-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16816333
Ver documentos Detalles de la promoción
Proteintech
10142-2-AP-150UL
150 μL
399.00€
150 microlitros
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 17-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

This antibody got two bands about 45 kDa and 50-55 kDa in western blotting, and the 50-55 kDa may be caused by phosphorylation.

cAMP is a signaling molecule important for a variety of cellular functions. cAMP exerts its effects by activating the cAMP-dependent protein kinase (AMPK), which transduces the signal through phosphorylation of different target proteins. The inactive holoenzyme of AMPK is a tetramer composed of two regulatory and two catalytic subunits. cAMP causes the dissociation of the inactive holoenzyme into a dimer of regulatory subunits bound to four cAMP and two free monomeric catalytic subunits. Four different regulatory subunits and three catalytic subunits of AMPK have been identified in humans. The protein encoded by this gene is one of the regulatory subunits. This subunit can be phosphorylated by the activated catalytic subunit. It may interact with various A-kinase anchoring proteins and determine the subcellular localization of AMPK.
Especificaciones

Especificaciones

PRKAR2A
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
P12367, P12368, P13861
19087, 29699, 5576
PRKAR2A Fusion Protein Ag0203
Primary
-20°C
PRKAR2A
0.31 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
PKR2, PRKAR2, PRKAR2A
Prkar2a
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documentos

Documentos

Product Certifications