missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PRKACA (PKA), His Tag Recombinant Protein
Human PRKACA (PKA), His Tag Recombinant Protein
Marca: Invitrogen™ PV4040
Detalles adicionales : Peso : 0.05250kg
Descripción
PRKACA (PKA) is a serine⁄threonine protein kinase that phosphorylates a broad spectrum of substrates. PRKACA regulates many cellular processes including cell growth, cell differentiation, ion-channel conductivity, gene transcription, and synaptic release of neurotransmitters.Especificaciones
NP_002721.1 | |
Protein Assays and Analysis | |
43.5kDa | |
1mg | |
For research use only. Not for use in diagnostic procedures | |
PRKACA (PKA) | |
PRKACA (PKA) was pre-diluted in enzyme dilution buffer and assayed in 50mM HEPES (pH 7.4), 24mM MgCl2, 240μM ATP, 180μg/mL kemptide peptide substrate (LRRASLG) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
PRKACA (PKA) Recombinant Human Protein | |
Human | |
E. coli | |
90% by SDS-PAGE |
0.33mg/mL | |
Frozen | |
PRKACA (PKA) Recombinant Human Protein | |
–80°C | |
1 Each | |
PRKACA (PKA) Recombinant Human Protein | |
PKA | |
4,800nmoles of phosphate transferred to Kemptide peptide substrate (LRRASLG) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.417μg/mL | |
His Tag | |
Frozen |
For Research Use Only. Not for use in diagnostic procedures.