missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PREPL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12478-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene belongs to the prolyl oligopeptidase subfamily of serine peptidases. Mutations in this gene have been associated with hypotonia-cystinuria syndrome, also known as the 2p21 deletion syndrome.Especificaciones
PREPL | |
Polyclonal | |
Unconjugated | |
PREPL | |
KIAA0436, PPCEL, PREPL, prolyl endopeptidase like, Prolylendopeptidase like | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
213760, 298771, 9581 | |
-20°C | |
Liquid |
Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q4J6C6, Q5HZA6, Q8C167 | |
PREPL | |
PREPL Fusion Protein Ag3196 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |