missing translation for 'onlineSavingsMsg'
Learn More

PPWD1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA553982

Detalles adicionales : Peso : 0.02000kg

Código de producto. 15701921

  • 514.00€ / 100 microlitros
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Immunogen sequence: GALFCSVGDD KAMKVFDVVN FDMINMLKLG YFPGQCEWIY CPGDAISSVA ASEKSTGKIF IYDGRGDNQP LHIFDKLHTS PLTQIRLNPV YKAVVS Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

PPWD1 Polyclonal antibody specifically detects PPWD1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Especificaciones

Especificaciones

PPWD1
Polyclonal
Unconjugated
PPWD1
4632422M10Rik; A330090G21Rik; KIAA0073; peptidylprolyl isomerase domain and WD repeat containing 1; peptidylprolyl isomerase domain and WD repeat-containing protein 1; PPWD1; RGD1310204; spliceosome-associated cyclophilin
Rabbit
Antigen affinity chromatography
RUO
23398
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96BP3
PPWD1
Recombinant protein corresponding to Human PPWD1
100 μL
Primary
Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

Research Use Only