missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PPP5C Recombinant Protein
Marca: Abnova™ H00005536-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Especificaciones
Antibody Production, ELISA, Protein Array, Western Blot | |
80.63 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25μg | |
-80°C | |
Recombinant |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
Human PPP5C Full-length ORF Recombinant Protein with GST-tag at N-terminal | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE stained with Coomassie Blue | |
Wheat Germ (in vitro) | |
Human | |
Solution |