missing translation for 'onlineSavingsMsg'
Learn More

PPP2R3A Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 1 publication

Marca:  Novus Biologicals NB100-93429

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215500

  • 369.32€ / 0.10 miligramo
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

PPP2R3A Polyclonal specifically detects PPP2R3A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Especificaciones

Especificaciones

PPP2R3A
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
PP2A subunit B isoform R3 isoform, PP2A subunit B isoforms B72/B130, PP2A subunit B isoforms B'-PR72/PR130, protein phosphatase 2 (formerly 2A), regulatory subunit B' (PR 72), alphaisoform and (PR 130), beta isoform, protein phosphatase 2 (formerly 2A), regulatory subunit B', alpha, protein phosphatase 2, regulatory subunit B', alpha, serine/threonine-protein phosphatase 2A regulatory subunit B' subunit alpha, subunit B, R3 isoform
Goat
Affinity Purified
RUO
5523
Human
IgG
Immunohistochemistry (Paraffin), ELISA
0.5 mg/mL
Immunohistochemistry-Paraffin, Peptide ELISA Detection limit 1:16000
Q06190
PPP2R3A
Peptide with sequence C-QKDVENDGPEPSD corresponding to internal region according to NP_002709.2, NP_871626.1.
0.1 mg
Primary
This antibody is expected to recognize both reported isoforms (NP_002709.2; NP_871626.1).
Store at -20C. Avoid freeze-thaw cycles.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only