missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PPP2R2A (Human) Recombinant Protein
Human PPP2R2A full-length ORF ( AAH41071, 1 a.a. - 447 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00005520-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MAGAGGGNDIQWCFSQVKGAVDDDVAEADIISTVEFNHSGELLATGDKGGRVVIFQQEQENKIQSHSRGEYNVYSTFQSHEPEFDYLKSLEIEEKINKIRWLPQKNAAQFLLSTNDKTIKLWKISERDKRPEGYNLKEEDGRYRDPTTVTTLRVPVFRPMDLMVEASPRRIFANAHTYHINSISINSDYETYLSADDLRINLWHLEITDRSFNIVDIKPANMEELTEVITAAEFHPNSCNTFVYSSSKGTIRLCDMRASALCDRHSKLFEEPEDPSNRSFFSEIISSISDVKFSHSGRYMMTRDYLSVKIWDLNMENRPVETYQVHEYLRSKLCSLYENDCIFDKFECCWNGSDSVVMTGSYNNFFRMFDRNTKRDITLEASRENNKPRTVLKPRKVCASGKRKKDEISVDSLDFNKKILHTAWHPKENIIAVATTNNLYIFQDKVN
Especificaciones
AAH41071 | |
protein phosphatase 2 (formerly 2A), regulatory subunit B, alpha isoform | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PPP2R2A | |
GST |
5520 | |
Wheat germ expression system | |
25 μg | |
B55-ALPHA, B55A, FLJ26613, MGC52248, PR52A, PR55A | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |