missing translation for 'onlineSavingsMsg'
Learn More

PPP1R37 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA559597

Detalles adicionales : Peso : 0.02000kg

Código de producto. 15747451

  • 510.00€ / 100 microlitros
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Immunogen sequence: TVDEVIGAYK QACQKLNCRQ IPKLLRQLQE FTDLGHRLDC LDLKGEKLDY KTCEALEEVF KRLQFKVVDL EQTNLDEDGA SALFDMIEYY ESATHL Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

PPP1R37 Polyclonal antibody specifically detects PPP1R37 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Especificaciones

Especificaciones

PPP1R37
Polyclonal
Unconjugated
PPP1R37
Gm158; KIAA1986; leucine rich repeat containing 68; leucine-rich repeat-containing protein 68; Lrrc68; PPP1R37; Protein phosphatase 1 regulatory subunit 37; protein phosphatase 1, regulatory subunit 37
Rabbit
Antigen affinity chromatography
RUO
284352
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O75864
PPP1R37
Recombinant protein corresponding to Human PPP1R37
100 μL
Primary
Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

Research Use Only