missing translation for 'onlineSavingsMsg'
Learn More

PPM1K Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA554136

Detalles adicionales : Peso : 0.02000kg

Código de producto. 15742071

  • 514.00€ / 100 microlitros
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Immunogen sequence: PETKRIKLHH ADDSFLVLTT DGINFMVNSQ EICDFVNQCH DPNEAAHAVT EQAIQYGTED NSTAVVVPFG AWGKYKNSEI NFSFS Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.

PPM1K Polyclonal antibody specifically detects PPM1K in Human samples. It is validated for Immunocytochemistry.
Especificaciones

Especificaciones

PPM1K
Polyclonal
Unconjugated
PPM1K
2900063A19Rik; A930026L03Rik; BDP; branched-chain alpha-ketoacid dehydrogenase phosphatase; hPTMP; im:6912645; MSUDMV; PP2C domain-containing protein phosphatase 1K; PP2Ckappa; PP2C-kappa; PP2C-like mitochondrial protein; PP2C-like protein; PP2CM; PP2C-type mitochondrial phosphoprotein phosphatase; PPM1K; PPM1K protein; protein phosphatase 1K (PP2C domain containing); protein phosphatase 1K, mitochondrial; Protein phosphatase 2C isoform kappa; protein phosphatase 2C kappa; protein phosphatase, Mg2+/Mn2+ dependent 1K; protein phosphatase, Mg2+/Mn2+ dependent, 1K; PTMP; RGD1308501; UG0882E07; zgc:113207
Rabbit
Antigen affinity chromatography
RUO
152926
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8N3J5
PPM1K
Recombinant protein corresponding to Human PPM1K
100 μL
Primary
Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

Research Use Only