missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PPM1A (Human) Recombinant Protein
Human PPM1A full-length ORF ( AAH26691, 1 a.a. - 382 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00005494-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MGAFLDKPKMEKHNAQGQGNGLRYGLSSMQGWRVEMEDAHTAVIGLPSGLESWSFFAVYDGHAGSQVAKYCCEHLLDHITNNQDFKGSAGAPSVENVKNGIRTGFLEIDEHMRVMSEKKHGADRSGSTAVGVLISPQHTYFINCGDSRGLLCRNRKVHFFTQDHKPSNPLEKERIQNAGGSVMIQRVNGSLAVSRALGDFDYKCVHGKGPTEQLVSPEPEVHDIERSEEDDQFIILACDGIWDVMGNEELCDFVRSRLEVTDDLEKVCNEVVDTCLYKGSRDNMSVILICFPNAPKVSPEAVKKEAELDKYLECRVEEIIKKQGEGVPDLVHVMRTLASENIPSLPPGGELASKRNVIEAVYNRLNPYKNDDTDSTSTDDMW
Especificaciones
AAH26691 | |
protein phosphatase 1A (formerly 2C), magnesium-dependent, alpha isoform | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PPM1A | |
GST |
5494 | |
Wheat germ expression system | |
10 μg | |
FLJ42306, MGC9201, PP2C-ALPHA, PP2CA | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |