Learn More
IP6 Kinase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA558765
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: CLENSEEVSQ PCQGVSVEVG KLVHKFHVGV GSLVQETLVE VGSPAEEIPE EVIQPYQEFS VEVGRLAQET SAINLLSQGI PEIDKPS Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 57%.
This gene encodes a dual functional inositol kinase. The encoded enzyme converts inositol hexakisphosphate to diphosphoinositol pentakisphosphate and diphosphoinositol pentakisphosphate to bis-diphosphoinositol tetrakisphosphate. This protein may be important for intracellular signaling pathways. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 15.Especificaciones
IP6 Kinase | |
Polyclonal | |
Unconjugated | |
PPIP5K1 | |
diphosphoinositol pentakisphosphate kinase 1; HISPPD2A; histidine acid phosphatase domain containing 2A; histidine acid phosphatase domain-containing protein 2A; hsVIP1; inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 1; Inositol pyrophosphate synthase 1; inositol-hexakisphosphate kinase; insP6 and PP-IP5 kinase 1; IP6 kinase; IP6K; IPS1; KIAA0377; PPIP5K1; VIP1; VIP1 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9677 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6PFW1 | |
PPIP5K1 | |
Recombinant protein corresponding to Human PPIP5K1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only