missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPIF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18466-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody is immunized with full length gene PPIF and absorbed by PPIA. So this antibody is specific to PPIF.
The protein encoded by this gene is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. This protein is part of the mitochondrial permeability transition pore in the inner mitochondrial membrane. Activation of this pore is thought to be involved in the induction of apoptotic and necrotic cell death.Especificaciones
PPIF | |
Polyclonal | |
Unconjugated | |
PPIF | |
Cyclophilin F, Cyp D, CYP3, peptidylprolyl isomerase F, PPIase F, PPIF, Rotamase F | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10105, 105675, 282819 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P29117, P30405, Q99KR7 | |
PPIF | |
PPIF-Specific Fusion Protein Ag13151 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |