Learn More
PPFIBP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA551663
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: DAQGFSDLEK SPSPTPVMGS PSCDPFNTSV PEEFHTTILQ VSIPSLLPAT VSMETSEKSK LTPKPETSFE ENDGNIILGA TVDTQLCDKL LTSSLQKSSS LGNLKKETSD GEKETIQKTS EDRA Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 67%.
PPFIBP1 Polyclonal antibody specifically detects PPFIBP1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
PPFIBP1 | |
Polyclonal | |
Unconjugated | |
PPFIBP1 | |
4632409B19Rik; AW214454; AW261454; hSGT2; hSgt2p; KIAA1230; L2; liprin beta 1; liprin related protein; liprin-beta 1; liprin-beta-1; PPFIA binding protein 1; Ppfibp1; Protein tyrosine phosphatase receptor type f polypeptide-interacting protein-binding protein 1; protein-tyrosine phosphatase receptor-type f polypeptide-interacting protein-binding protein 1; PTPRF interacting protein, binding protein 1 (liprin beta 1); PTPRF-interacting protein-binding protein 1; SGT2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8496 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86W92 | |
PPFIBP1 | |
Recombinant protein corresponding to Human PPFIBP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only