missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16662-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein which interacts with the carboxy-terminus of transcription initiation factor TFIIF, a transcription factor which regulates elongation as well as initiation by RNA polymerase II. The protein may also represent a component of an RNA polymerase II holoenzyme complex. Alternative splicing of this gene results in two transcript variants encoding 2 different isoforms.Especificaciones
PPA2 | |
Polyclonal | |
Unconjugated | |
PPA2 | |
HSPC124, PPA2, PPase 2, pyrophosphatase (inorganic) 2, Pyrophosphatase SID6 306, SID6 306 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27068, 310856, 74776 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q91VM9, Q9H2U2 | |
Ppa2 | |
PPA2 Fusion Protein Ag10138 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |