Learn More
PPA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA554146
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: NAEFKDKDFA IDIIKSTHDH WKALVTKKTN GKGISCMNTT LSESPFKCDP DAARAIVDAL PPPCESACTV PTDVDKWFHH Q Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 86%.
PPA1 Polyclonal antibody specifically detects PPA1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
PPA1 | |
Polyclonal | |
Unconjugated | |
Ppa1 | |
2010317E03Rik; cytosolic inorganic pyrophosphatase; diphosphate phosphohydrolase; epididymis secretory sperm binding protein Li 66p; HEL-S-66p; inorganic diphosphatase; inorganic diphosphatase 1; inorganic pyrophosphatase; inorganic pyrophosphatase 1; IOPPP; Pp; PP1; Ppa1; PPase; Pyp; pyrophosphatase (inorganic) 1; pyrophosphatase 1; pyrophosphate phospho-hydrolase; RP11-367H5.1; SID6-8061 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5464 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15181 | |
Ppa1 | |
Recombinant protein corresponding to Human PPA1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only