Learn More
Fisherbrand™ PowerPro 500
Product Code. 15838481 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Display:
2.4 in. TFT-LCD
Unit Size:
Each
Product Code. Display Unit Size
15838481 2.4 in. TFT-LCD Each
1 options
This item is not returnable. View return policy
Product Code. 15838481
  • ONLINE EXCLUSIVE PRICE
    1845.00€
    889.19€ / paquete de 1
    Ahorro 955.81€
    51% Descuento
Fecha estimada de envoi: 03-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Fisherbrand™ PowerPro 500 is an excellent general-purpose power supply that fits the broadest range of electrophoresis applications, and can run as many as four units under a constant or programmable setting.
  • Perfect for horizontal or vertical electrophoresis
  • Rigorous in-built safety mechanisms
  • Dual voltage compatibility
  • 5 pairs of outlets
  • It has the capacity to store up to 30 programmed files, each with 6 steps.
  • Perfect for techniques that benefit from subtle and gradual stepwise changes in the electrical parameters as the run progresses, such as DGGE and large format vertical PAGE using Cleaver Scientific VS20-DGGE and WAVE systems.
  • Built-in pre-set protocols from running each of Cleaver Scientific’s electrophoresis systems
  • Users can create and save protocols for their own systems.
  • A 2.4” TFT-LCD Screen displays the current running parameters and allows the user to browse pre-set and saved protocols.
  • Can be operated at constant voltage, constant current or constant power. The convenient pause/resume key allows any run to be temporarily halted for inspection of the gel and restarted from the same position.
Specifications
For Use With (Application) Broad range of electrophoresis applications
Timer Constant mode: 9999 (min.) with alarm Programmable mode: 999 (min.) with alarm
Display 2.4 in. TFT-LCD
Voltage 100 to 240 V rated, 5 to 500 V output, 1 V increments
Wattage 300 w output, 1 w increments
Amperage 1 to 800 mA output, 1 mA increments