Learn More
Aalborg™ Power Supply
Product Code. 11787959
Change view
Click to view available options
For Use With (Equipment):
Digital Flowmeter
Gas Mass Flow Controller
Gas Mass Flowmeter
Type:
DFM Adapter
GFC Adapter
Power Supply
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Type Unit Size
11787959 Gas Mass Flow Controller GFC Adapter Each
11787709 Gas Mass Flowmeter Power Supply Each
11797619 Digital Flowmeter DFM Adapter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11787959 Supplier Aalborg™ Supplier No. PSGFC230EU2
  • ONLINE EXCLUSIVE PRICE
    126.83€
    75.48€ / paquete de 1
    Ahorro 51.35€
    40% Descuento
 Restricted Item
Fecha estimada de envoi: 11-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Aalborg™ Power Supply is a component that supplies power to at least one electric load.
  • Accessory for DFM, GFM, or GFC
  • 230VAC Continential Europe Plug 12VDC
  • Power supply
Specifications
Voltage 230 V
For Use With (Equipment) Gas Mass Flow Controller
Type GFC Adapter
Electrical Requirements 230VAC, 12VDC