Learn More
Uvex™ Power Protect V1000 Safety Glove
The Uvex power protect V1000 protective glove offers safe protection against electrical voltages up to 1000 V.
Marca: Uvex™ 6084010
Detalles adicionales : Peso : 0.28400kg
Descripción
- The glove's high level of flexibility offers pleasant wearer comfort and an excellent tactile feel.
- Anatomical fit
- Flexible, with a good grip feel even at low temperatures
- Flexible at cold temperatures
- Mineral oil-resistant
Especificaciones
EN 60903: 2003 IEC 60903: 2014 (Class 0/RC) | |
Red | |
Latex | |
10 |