Learn More
1000ml Potassium Hydroxide, Alcoholic solution EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA
Product Code. 12990874 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
Product Code. Quantity unitSize
12990874 1000 mL 1000mL
1 options
This item is not returnable. View return policy
Product Code. 12990874 Supplier Fisher Chemical Supplier No. VL 041 1000
This item is not returnable. View return policy
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Specifications
Concentration 0.01 M
Quantity 1000 mL
compliance-icons
Product Identifier
  • Potassium hydroxide,Alcoholic. 0.01M Volumetric Solution European Pharmacopoeia Ref: 3009000
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501-Dispose of contents/container to .