missing translation for 'onlineSavingsMsg'
Learn More

Fluoruro de potasio dihidrato, +98,5 %, Thermo Scientific Chemicals

Catalog Number p-7048819
Click to view available options
Cantidad:
25 g
250 g
1 kg
Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: es posible que la compra de este producto esté sujeta a restricciones y necesitemos solicitarle documentación adicional.
13455-21-5
FH4KO2
94.13
MFCD00149929
MZFXHSQFQLKKFD-UHFFFAOYSA-M
potassium fluoride dihydrate, unii-95r1d12heh, potassium dihydrate fluoride, potassium fluoride dihydrate, 98.5+%, potassium fluoride dihydrate, puratronic, potassium fluoride, dihydrate, potassium fluoride kf , dihydrate, acmc-1byyf, ksc497c4l, potassiumfluoridedihydrate
23708904
O.O.[F-].[K+]
Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: es posible que la compra de este producto esté sujeta a restricciones y necesitemos solicitarle documentación adicional.
13455-21-5
FH4KO2
94.13
MFCD00149929
MZFXHSQFQLKKFD-UHFFFAOYSA-M
potassium fluoride dihydrate, unii-95r1d12heh, potassium dihydrate fluoride, potassium fluoride dihydrate, 98.5+%, potassium fluoride dihydrate, puratronic, potassium fluoride, dihydrate, potassium fluoride kf , dihydrate, acmc-1byyf, ksc497c4l, potassiumfluoridedihydrate
23708904
O.O.[F-].[K+]

Potassium fluoride dihydrate is used as a stationary phase in gas chromatography. It is also used for the conversion of chlorocarbons into fluorocarbons through Finkelstein reaction. Further, it is used as a fluxing agent of silver brazing and metallurgical processing. It serves as an analytical reagent, complexing agent and masking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Potassium fluoride dihydrate
Punto de fusión 41°C
Forma física cristalino/a/s
Cantidad 25 g
Número UN UN1812
Índice Merck 14,7632
Información de solubilidad Freely soluble in waterSoluble in water; Insoluble in ethanol.
Nombre IUPAC potassium dihydrate fluoride
Formula Weight (peso de la fórmula) 94.13 (58.10 Anhydrous)
Porcentaje de pureza ≥98.5%
Olor Odorless
Índice de refracción 1.352
Sensibilidad Hygroscopic
Densidad 2.5 g/mL
Mostrar más Mostrar menos

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'