missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromate, ≥99.8%, ACS Reagent, Honeywell™ Fluka™

Catalog Number 15613150
missing translation for 'orderingAttributeHoverText'
Cantidad:
5 g
100 g
500 g
Envase:
Glass Bottle
This item is not returnable. View return policy
7758-01-2
BrKO3
167.00
MFCD00011359
OCATYIAKPYKMPG-UHFFFAOYSA-M
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
23673461
CHEBI:38211
potassium;bromate
[O-]Br(=O)=O.[K+]
This item is not returnable. View return policy
7758-01-2
BrKO3
167.00
MFCD00011359
OCATYIAKPYKMPG-UHFFFAOYSA-M
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
23673461
CHEBI:38211
potassium;bromate
[O-]Br(=O)=O.[K+]
Nombre del producto químico o material Potassium Bromate
Punto de fusión 42° to 487deg;C
Cantidad 100 g
Fórmula lineal KBrO3
Número UN UN1484
Formula Weight (peso de la fórmula) 167g/mol
Porcentaje de pureza ≥99.8%
Grado ACS Reagent
Envase Glass Bottle
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'