missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ POP-7™ Polymer For 3130/3130xL Genetic Analyzers

POP-7™ Polymer is a separation matrix for performing DNA sequencing and fragment analysis on the Applied Biosystems 3130 and the Applied Biosystems 3130xl Genetic Analyzers.

Marca:  Applied Biosystems™ 4363785

Detalles adicionales : Peso : 0.03000kg

 Ver más versiones de este producto

Código de producto. 15843550

  • 295.00€ / 3.50 ml


En stock 685
Añadir a la cesta

Descripción

Descripción

  • Easy to use: Pre-formulated, which not only saves time but also saves reproducibility by ensuring consistent formulation
  • Robust formulation: Formulated for sequencing and fragment analysis applications
  • Replenishable: Allow capillaries to be used multiple times

Order Info

Each

Especificaciones

Especificaciones

98.5% upto 500 bases (for 36cm capillary), 98.5% upto 950 bases (for 80cm capillary) accuracy, Stability on instrument 7 days, Up to 250 samples/3.5mL
Particulate-Free Viscous Solution
Liquid
Clear
Videos

For research use only