Learn More
Applied Biosystems™ POP-6™ Polymer For 3130/3100xl Genetic Analyzers
The POP-6™ Polymer is a separation matrix for performing standard and rapid DNA sequencing on the Applied Biosystems 3130 and the Applied Biosystems 3130xl Genetic Analyzers.
Marca:  Applied Biosystems™ 4363783
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15819816
  • 311.00€ / cada uno
Stock EU 129
Añadir a la cesta
Descripción
Descripción
  • Easy to use: Pre-formulated, which not only saves time but also saves reproducibility by ensuring consistent formulation
  • Robust formulation: Formulated for sequencing and fragment analysis applications
  • Replenishable: Allow capillaries to be used multiple times
Order Info
Each
Especificaciones
Especificaciones
POP-6™ Polymer For 3130/3130xL Genetic Analyzers
3130 Genetic Analyzer, 3130xL Genetic Analyzer
Contains Enough Polymer for 960 Samples
1 Hour For 36cm capillary, 2 Hour 45 Minutes For 50cm capillary
Rapid Sequencing, Standard Sequencing
POP-6™ Polymer
Store at 2 to 8°C
3500μL
Videos
For research use only