missing translation for 'onlineSavingsMsg'
Learn More

Poliestireno estándar, P.M. 30000, Pm/Nm 1,06, Thermo Scientific™

Catalog Number p-4824884
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
9003-53-6
(C8H8)n
104.15
MFCD00084450
PPBRXRYQALVLMV-UHFFFAOYSA-N
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
7501
CHEBI:27452
*-CC(-*)C1=CC=CC=C1
Este artículo no se puede devolver. Vea la política de devoluciones
9003-53-6
(C8H8)n
104.15
MFCD00084450
PPBRXRYQALVLMV-UHFFFAOYSA-N
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
7501
CHEBI:27452
*-CC(-*)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Polystyrene standard
Color Blanco
Nombre de nota M.W. 30, 000, Mw/Mn 1.06
Forma física Polvo
Cantidad 1 g
Nombre IUPAC estireno
Olor Odorless
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'