Learn More
Kartell™ Polystyrene Cuvettes
Achieve more constant sampling conditions when handling samples in a variety of applications. Kartell™ Polystyrene Cuvettes are ideal for spectrophotometry and when determining coagulation time.
Marca:  Kartell™ 0263100
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 12016992
  • 48.20€ / paquete de 1000
Stock Local 8
Añadir a la cesta
Especificaciones
Especificaciones
Cuvettes with Metallic Mixing Balls
11.25mm Diameter
Amelung™
1000 Metallic Mixer
1 mL
CE IVD
Ideal for spectrophotometry application
22.8mm
1000Pack
Polystyrene