Learn More
Kartell™ Micropipette Filter Tip
Incase of over-aspiration these tips feature orifices that are sized to completely empty the tip. Kartell™ Micropipette Filter Tip eliminates cross-contamination of samples and is compatable with most micropipette brands.
Marca:  Kartell™ 0963700
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 12583247
  • 144.00€ / paquete de 960
Fecha estimada de envío
16-06-2020
Añadir a la cesta
Descripción
Descripción
  • Includes a filter
  • DNA-free
  • RNA-free
  • Cadmium-free
  • Apyrogenic
  • Sterilized by gamma radiation
  • Autoclavable at 121°C for 20 minutes
  • Designed for use in molecular biology, cell cultures, bacteriology, virology, immunology and radioimmunology
Especificaciones
Especificaciones
Rack ster.
Filtered
Natural
PP (Polypropylene)
Molecular biology, cell cultures, bacteriology, and more
Tip
5 to 300μL
Sterile
Yes
CE/IVD
Eppendorf™, Gilson™, Biohit™, Hamilton™
Gamma Radiation