Learn More
Kartell™ Micropipette Filter Tip
Incase of over-aspiration these tips feature orifices that are sized to completely empty the tip. Kartell™ Micropipette Filter Tip eliminates cross-contamination of samples and is compatable with most micropipette brands.
Marca:  Kartell™ 0963900
Detalles adicionales : Peso : 0.10000kg
 Ver más versiones de este producto
Código de producto. 10796722
  • 220.00€ / paquete de 960
Consúltenos : 902 239 303 
Añadir a la cesta
Especificaciones
Especificaciones
Rack ster.
Filtered
Natural
PP (Polypropylene)
Molecular biology, cell cultures, bacteriology, and more
Tip
2 to 20μL
Sterile
Yes
CE/IVD
Kartell™ PL100, Gilson™
Gamma Radiation