Learn More
Brady™ Polypropylene Extra Cleats for 480/600V Lockout
Product Code. 15633507 Shop All Brady Products
Change view
Click to view available options
Color:
Red
Unit Size:
Pack of 6
Product Code. Color Unit Size
15633507 Red Pack of 6
1 options
This item is not returnable. View return policy
Product Code. 15633507 Supplier Brady™ Supplier No. 065406
  • ONLINE EXCLUSIVE PRICE
    21.10€
    18.40€ / paquete de 6
    Ahorro 2.70€
    12% Descuento
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Cleats easily clip onto the bottom of clamp-on breaker lockout devices.
  • Assures positive restraint for breakers with long, sliding switch throws
  • Polypropylene
Specifications
Product Type Extra Cleats
Alternate Part Number LOCKOUT CLEAT 480/600 VOLT GENERAL USE
Color Red
Description Extra Cleats for 480/600V Lockout
For Use With (Equipment) For 480/600V Lockout
Quantity 6 Pk.