Learn More
Cole-Parmer™ Polypropylene Cable Ties
Product Code. 11723391 Shop All Cole-Parmer Products
Change view
Click to view available options
Color:
Black
White
Length (Metric):
10 cm
20 cm
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Length (Metric) Unit Size
11723391 White 10 cm Pack of 1000
11733391 Black 20 cm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11723391 Supplier Cole-Parmer™ Supplier No. 0683085
  • ONLINE EXCLUSIVE PRICE
    332.00€
    234.79€ / paquete de 1000
    Ahorro 97.21€
    29% Descuento
Fecha estimada de envoi: 18-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Keep items bundled during autoclaving with these ties. Cole-Parmer™ Polypropylene Cable Ties are ideal for tubing bundles.
  • Polypropylene (PP) cable ties resist a number of chemicals
  • UV-resistant
  • Sold in packs of 1000
Specifications
Type Cable Tie
Material Polypropylene
Color White
Length (Metric) 10 cm
Quantity 1000/Pk.
For Use With (Application) Ideal for tubing bundles