Learn More
Kimberly-Clark™ KLEENGUARD™ A80 Chemical Permeation and Jet Liquid Protection Coveralls
Specially designed for increased protection against chemical splashes. Kimberly-Clark™ KLEENGUARD™ A80 Chemical Permeation and Jet Liquid Protection Coveralls have welded seams that improve safety in high-risk areas creating a jet-tight barrier against harmful substances.
Marca:  Kimberly-Clark™ 96550
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 12611396
  • 364.14€ / paquete de 10
Fecha estimada de envío
11-09-2019
Añadir a la cesta
Descripción
Descripción
  • Approved as Complex Design (Category 3) equipment - offering protection to the levels specified for Type 3 and 5 by CEN
  • Highly visible, single-use protective clothing to improve safety and comfort in high-risk areas - creates a high pressure jet tight barrier against harmful chemical splashes, liquids or dust particles
  • Suitable for use as protection against Infective Agents and Biohazards - EN 14126
  • Suitable for use as protection against solid particles and particulate radioactive contamination - EN1073-2
  • Treated to be antistatic to EN 1149-5
  • Ideal for working with hazardous chemicals - protecting against liquid, aerosol and dry chemical splashes, sprays, high pressure jets, fibres and dust when used for heavy industrial clean-up, maintenance, tank cleaning, petrochemical and chemical handling
  • Perfect for environments where high levels of body protection are vital, such as chemical and manufacturing industries
  • Durable SMS fabric with additional treatment for increased protection against chemical splashes
  • Offers complete freedom of movement with comfortable body design
Especificaciones
Especificaciones
Orange
3X-Large
A80 Coveralls