Learn More
Kartell™ Plastilab™ Polypropylene 4-Way Connectors
For use with plastic or rubber tubing.
Marca: Kartell™ 7141300
Detalles adicionales : Peso : 0.02340kg
Descripción
- Sturdy shape provides strength
- Chemical and thermal resistant
Especificaciones
CE IVD | |
Autoclavable | |
10/Pk. | |
4-way cross |
10 mm | |
Polypropylene | |
7mm Bore | |
Natural |