Learn More
Kartell™ Plastilab™ Polypropylene 4-Way Connectors
For use with plastic or rubber tubing.
Marca: Kartell™ 7141100
Detalles adicionales : Peso : 0.00900kg
Descripción
- Sturdy shape provides strength
- Chemical and thermal resistant
Especificaciones
CE IVD | |
Autoclavable | |
10/Pk. | |
4-way cross |
6 mm | |
Polypropylene | |
2.7mm Bore | |
Natural |