Learn More
Polietilenglicol monometileter, 350, Thermo Scientific Chemicals
Catalog Number 11314328
Click to view available options
Cantidad:
100 g
500 g
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:21615
9004-74-4
(C2H4O)nCH4O
76.10
MFCD00084416
XNWFRZJHXBZDAG-UHFFFAOYSA-N
methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
8019
CHEBI:46790
2-methoxyethan-1-ol
COCCO
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:21615
9004-74-4
(C2H4O)nCH4O
76.10
MFCD00084416
XNWFRZJHXBZDAG-UHFFFAOYSA-N
methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
8019
CHEBI:46790
2-methoxyethan-1-ol
COCCO
Polyethylene glycol monomethylether is used as enteric release coatings. It is also used for a series of polycarboxylate water reducing agent. It acts as a solvent for brake fluids. Further, it is used in the building materials industry and raw material for cement water reducing agent and strengthening agent. In addition to this, it is used in surfactants, polyester and polyurethane based paints.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Polyethylene glycol monomethylether, 350
Color De incoloro a amarillo
Forma física Líquido
Densidad 1.09 g/mL
Información de solubilidad Partially miscible with water.
Cantidad 500 g
Temperatura de inflamación >110°C (230°F)