Learn More
Alfa Aesar™ Polyethylene glycol methyl ether methacrylate, M.W. 1,000
Marca:  Alfa Aesar™ 046991.06
Detalles adicionales : CAS : 26915-72-0 Peso : 0.00500kg
Cantidad. 5g
 Ver más versiones de este producto
Código de producto. 15497355
  • 360.00€ / 5 g
Stock EU 1
Añadir a la cesta
Especificaciones
26915-72-0
276.329
KRCGBOKYIUDIFY-UHFFFAOYSA-N
93675
CC(=C)C(=O)OCCOCCOCCOCCOC
C13H24O6
MFCD00241432
2-methoxyethyl methacrylate, beta-methoxyethyl methacrylate, methacrylic acid, 2-methoxyethyl ester, 2-propenoic acid, 2-methyl-, 2-methoxyethyl ester, poly ethylene glycol methyl ether methacrylate, pegmem, peg-mem, acmc-209oay, methoxyethyl methacrylate, 2-methoxyethylmethacrylate
2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethyl 2-methylprop-2-enoate
Especificaciones
Polyethylene glycol methyl ether methacrylate
26915-72-0
MFCD00241432
KRCGBOKYIUDIFY-UHFFFAOYSA-N
2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethyl 2-methylprop-2-enoate
93675
White
5g
M.W. 1,000
C13H24O6
2-methoxyethyl methacrylate, beta-methoxyethyl methacrylate, methacrylic acid, 2-methoxyethyl ester, 2-propenoic acid, 2-methyl-, 2-methoxyethyl ester, poly ethylene glycol methyl ether methacrylate, pegmem, peg-mem, acmc-209oay, methoxyethyl methacrylate, 2-methoxyethylmethacrylate
CC(=C)C(=O)OCCOCCOCCOCCOC
276.329
Granular Solid
33°C to 38°C
Videos
SDS Documentos