Learn More
Alfa Aesar™ Polyethylene glycol dimethanesulfonate, M.W. 3,400
Marca:  Alfa Aesar™ 046806.06
Detalles adicionales : CAS : 52410-74-9 Peso : 0.00500kg
Cantidad. 5g
 Ver más versiones de este producto
Código de producto. 15492767
  • 382.00€ / 5 g
Fecha de envío estimada
del proveedor 16-10-2019
Añadir a la cesta
Especificaciones
52410-74-9
218.238
QSQFARNGNIZGAW-UHFFFAOYSA-N
20796
CS(=O)(=O)OCCOS(=O)(=O)C
C4H10O6S2
MFCD03100947
ethylene dimethanesulfonate, dmse, 1,2-bis mesyloxy ethane, ethylene methanesulfonate, ethylene glycol dimesylate, ethane dimethane sulphonate, 1,2-ethanediyl dimethanesulfonate, dimethylene dimethanesulfonate, ethylene bis methanesulfonate, ethane dimethanesulphonate
2-methylsulfonyloxyethyl methanesulfonate
Especificaciones
Polyethylene glycol dimethanesulfonate
52410-74-9
MFCD03100947
QSQFARNGNIZGAW-UHFFFAOYSA-N
2-methylsulfonyloxyethyl methanesulfonate
20796
White
M.W. 3,400
C4H10O6S2
ethylene dimethanesulfonate, dmse, 1,2-bis mesyloxy ethane, ethylene methanesulfonate, ethylene glycol dimesylate, ethane dimethane sulphonate, 1,2-ethanediyl dimethanesulfonate, dimethylene dimethanesulfonate, ethylene bis methanesulfonate, ethane dimethanesulphonate
CS(=O)(=O)OCCOS(=O)(=O)C
218.238
Granular Solid
5g
Videos
SDS Documentos