Learn More
Cytiva Polyetheretherketone Solenoid Valve PSV-50
Product Code. 11668883 Shop All Cytiva Products
Change view
Click to view available options
Operating Pressure:
2 bar
Unit Size:
Each
Product Code. Operating Pressure Unit Size
11668883 2 bar Each
1 options
This item is not returnable. View return policy
Product Code. 11668883 Supplier Cytiva Supplier No. 19199401
Fecha estimada de envoi: 07-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Excellent solvent resistance
Solenoid valves PSV-50 and the three-way valve may be operated by most fraction collectors. Both have pressure limits of 2 bar (0.2 MPa, 29 psi) and excellent solvent resistance. The PSV-50 is used for gradient formation with HiLoad Pump P-50 and flow diversion with most fraction collectors. Long life and zero dead volume make the three-way valve an excellent choice for fraction collector flow diversion in SuperFrac.
  • A range of manual and automated valves to meet a variety of performance requirements.
  • Designed for biocompatibility and years of reliable service, each may be inserted into the flow path without causing unnecessary mixing or sample dilution.
TRUSTED_SUSTAINABILITY
Specifications
Material Ceramic, PTFE, ECTFE
Model PSV-50
Operating Pressure 2 bar
Type Solenoid Valve PSV-50