missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polidimetilsiloxano, trimetilsiloxio terminado, P.M. 770, Thermo Scientific Chemicals
Click to view available options
Cantidad:
50 mL
250 mL
1 L
Descripción
Polydimethylsiloxane, trimethylsiloxy is used in coating Auxiliary Agents. It is also used as Pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Nombre del producto químico o material | Polydimethylsiloxane, trimethylsiloxy terminated |
Color | Incolora |
Nombre de nota | M.W. 770 |
Forma física | Líquido |
Punto de ebullición | >205°C |
Punto de fusión | -40°C |
Densidad | 0.918 g/mL |
Información de solubilidad | Soluble in aromatic hydrocarbon,partially soluble in aliphatic and insoluble in water. |
Cantidad | 50 mL |
Temperatura de inflamación | 135°C (275°F) |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'