Learn More
Polidimetilsiloxano, trimetilsiloxio terminado, P.M. 4000, Thermo Scientific Chemicals
Descripción
Polydimethylsiloxane, trimethylsiloxy terminated is widely used as a common surfactant. It is an active component of defoamers which suppress the formation of foams and as a critical ingredient in water-repelling coatings as an active silicone fluid. It finds application in optic telecommunications, biomedical research, elasticity of polymer networks, adhesive materials and in skin-moisturizing lotions. Furthermore, it is used as a filler fluid in breast implants and as a sorbent for the analysis of headspace (dissolved gas analysis) of food.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Nombre del producto químico o material | Polydimethylsiloxane, trimethylsiloxy terminated |
Color | Incolora |
Nombre de nota | M.W. 4,000 |
Forma física | Líquido viscoso |
Punto de ebullición | >225°C |
Punto de fusión | <-40°C |
Densidad | 0.960 g/mL |
Información de solubilidad | Slightly miscible with water. Immiscible with methanol and ethylene glycol. |
Cantidad | 50 mL |
Temperatura de inflamación | 285°C (545°F) |
Mostrar más |
RUO – Research Use Only