missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polycystin 2 Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19126-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the polycystin protein family. The encoded protein contains multiple transmembrane domains, and cytoplasmic N- and C-termini. The protein may be an integral membrane protein involved in cell-cell/matrix interactions. The encoded protein may function in renal tubular development, morphology, and function, and may modulate intracellular calcium homoeostasis and other signal transduction pathways. This protein interacts with polycystin 1 to produce cation-permeable currents. Mutations in this gene have been associated with autosomal domit polycystic kidney disease.Especificaciones
Polycystin 2 | |
Polyclonal | |
Unconjugated | |
PKD2 | |
APKD2, PC2, PKD2, PKD4, Polycystin 2, Polycystwin, R48321 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18764, 487852, 498328, 5311 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35245, Q13563 | |
Pkd2 | |
Peptide | |
20 μL | |
Primary | |
Rat, Mouse, Canine, Human | |
Antibody | |
IgG |