missing translation for 'onlineSavingsMsg'
Learn More

Poli(2-etil-2-oxazolina), P.M. 200000, Thermo Scientific Chemicals

Catalog Number p-7052016
Click to view available options
Cantidad:
100 g
500 g
2.5 kg
Este artículo no se puede devolver. Vea la política de devoluciones
25805-17-8
(C5H9NO)n
99.13
MFCD00134377
NYEZZYQZRQDLEH-UHFFFAOYSA-N
2-ethyl-2-oxazoline, 2-ethyloxazoline, 2-ethyl-4,5-dihydrooxazole, oxazole, 2-ethyl-4,5-dihydro, 2-oxazoline, 2-ethyl, unii-b8cd92t4p5, unii-jm7uh9q3yn, unii-hnx7574gtx, unii-s5dpg6k67i, ethyl oxazoline
66412
2-ethyl-4,5-dihydro-1,3-oxazole
CCC(=O)N(-*)CC-*
Este artículo no se puede devolver. Vea la política de devoluciones
25805-17-8
(C5H9NO)n
99.13
MFCD00134377
NYEZZYQZRQDLEH-UHFFFAOYSA-N
2-ethyl-2-oxazoline, 2-ethyloxazoline, 2-ethyl-4,5-dihydrooxazole, oxazole, 2-ethyl-4,5-dihydro, 2-oxazoline, 2-ethyl, unii-b8cd92t4p5, unii-jm7uh9q3yn, unii-hnx7574gtx, unii-s5dpg6k67i, ethyl oxazoline
66412
2-ethyl-4,5-dihydro-1,3-oxazole
CCC(=O)N(-*)CC-*

Poly(2-ethyl-2-oxazoline) shows the application in biocompatibility, avoiding accumulation in tissue, and rapid clearance from the bloodstream. It is also adapted to many different functions for both protein and small drug conjugation. It also acts as a perfect substitute for poly(vinyl alcohol) and poly(vinyl pyrrolidone). It is also used as an adhesion promoter in coatings and heat sealing. It behaves as a hydrophilic block and utilized in the synthesis of amphiphilic diblock copolymers by reacting with poly(1,3-trimethylene carbonate) (PTMC) as a hydrophobic block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Poly (2-ethyl-2-oxazoline)
Nombre de nota M.W. 200,000
Forma física Sólido blanquecino
Información de solubilidad Soluble in water and organic solvents.
Cantidad 100 g

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'