Learn More
POLR3F Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA561971
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: CELGISKVEL SMEDIETILN TLIYDGKVEM TIIAAKEGTV GSVDGHMKLY RAVNPIIPPT GLVRAPCGLC PVFDDCHEGG EISPSNCI Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.
The human POLR3F is a component of RNA III polymerase. RNA polymerase III transcribes many essential, small, noncoding RNAs, including the 5S rRNAs and tRNAs. While most pol III-transcribed genes are found scattered throughout the linear chromosome maps or in multiple linear clusters, there is increasing evidence that many of these genes prefer to be spatially clustered, often at or near the nucleolus. This association could create an environment that fosters the coregulation of transcription by pol III with transcription of the large ribosomal RNA repeats by RNA polymerase I (pol I) within the nucleolus. Given the high number of pol III-transcribed genes in all eukaryotic genomes, the spatial organization of these genes is likely to affect a large portion of the other genes in a genome. POLR3F has also been recently identified as an HIV dependency factor (HDF), suggesting that POLR3F may be an important drug target in HIV treatment. At least two isoforms of POLR3F are known to exist.Especificaciones
POLR3F | |
Polyclonal | |
Unconjugated | |
POLR3F | |
2810411G20Rik; 3010019O03Rik; 3110032A07Rik; DNA-directed RNA polymerase III subunit F; DNA-directed RNA polymerase III subunit RPC6; Polr3f; polymerase (RNA) III (DNA directed) polypeptide F; polymerase (RNA) III (DNA directed) polypeptide F, 39 kDa; polymerase (RNA) III subunit F; RNA polymerase III 39 kDa subunit; RNA polymerase III C39 subunit; RNA polymerase III subunit C6; RNA polymerase III subunit F; RPC39; RPC6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10621 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H1D9 | |
POLR3F | |
Recombinant protein corresponding to Human POLR3F | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only