Learn More
POLR2J2 Monoclonal Antibody (1B2), Invitrogen™
Mouse Monoclonal Antibody
Marca:  Invitrogen MA524531
Detalles adicionales : Peso : 0.01000kg
Código de producto. 15692355
  • 473.00€ / 100 microgramos
Fecha estimada de envío: 17-05-2024
para ver el stock
Descripción
Descripción
Peptide Sequence: MNAPPAFESF LLFEGEKITI NKDTKVPKAC LFTINKEDHT LGNIIKSQLL KDPQVLFAGY KVPHPLEHKI IIRVQTTPDY SPQEAFTNAI TDLISELSLL EERFRTCLLP LRLLP
POLR2J2 Monoclonal antibody specifically detects POLR2J2 in Human samples. It is validated for ELISA, Western Blot.
Especificaciones
Especificaciones
POLR2J2
Monoclonal
Unconjugated
POLR2J2
DNA directed RNA polymerase II polypeptide J-related; DNA-directed RNA polymerase II subunit J2; DNA-directed RNA polymerase II subunit RPB11-b1; HRPB11B; POLR2J2; polymerase (RNA) II (DNA directed) polypeptide J2; polymerase (RNA) II subunit J2; RNA polymerase II subunit B11-b1; RNA polymerase II subunit J2; RPB11b1
Mouse
Protein A
RUO
246721
-20° C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1B2
PBS with no preservative; pH 7.4
Q9GZM3
POLR2J2
POLR2J2 (NP_116581.3, 1 a.a. ∽ 115 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ