Learn More
POLA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA551784
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: VDLEPMAAKA WDKESEPAEE VKQEADSGKG TVSYLGSFLP DVSCWDIDQE GDSSFSVQEV QVDSSHLPLV KGADEEQVFH FYWLDAYEDQ YNQPGVVFLF GKVWIESAET HVSCCVMVK Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 76%.
POLA1 Polyclonal antibody specifically detects POLA1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Especificaciones
POLA1 | |
Polyclonal | |
Unconjugated | |
POLA1 | |
AW321876; DNA pol alpha; DNA polymerase alpha 1 catalytic subunit; DNA polymerase alpha 1, 180 kDa; DNA polymerase alpha 1, catalytic subunit; DNA polymerase alpha catalytic subunit; DNA polymerase alpha catalytic subunit p180; DNA polymerase alpha p180 subunit; DNA polymerase alpha subunit I; G7a/Bat6Hom; NSX; p180; Pola; Pola1; polymerase (DNA directed), alpha 1; polymerase (DNA directed), alpha 1, catalytic subunit; polymerase (DNA) alpha 1, catalytic subunit; polymerase (DNA-directed), alpha (70kD); polymerase, alpha 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5422 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P09884 | |
POLA1 | |
Recombinant protein corresponding to Human POLA1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only